Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-bromooctanoate, 98+%

Manufacturer:  Alfa Aesar™ B2189622

Quantity 100g

 View more versions of this product

Catalog No. AAB2189622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

5445-29-4
251.17
1767535
Ethyl 2-bromooctanoate
C10H19BrO2
MFCD00042603
2-Bromooctanoic acid ethyl ester

Specifications

5445-29-4
C10H19BrO2
106°C (222°F)
MFCD00042603
2-Bromooctanoic acid ethyl ester
≥98%
137° to 140°C (25mmHg)
100g
251.17
1767535
Ethyl 2-bromooctanoate

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit