Alfa Aesar™ Ethyl 2-bromothiazole-5-carboxylate, 98%

Manufacturer: Alfa Aesar™ H5510903

Quantity 1g

 View more versions of this product

Catalog No. AAH5510903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41731-83-3
    Molecular Formula C6H6BrNO2S
    Formula Weight 236.08
    MDL Number MFCD00463837
    Synonym 2-Bromothiazole-5-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 2-bromothiazole-5-carboxylate

    Specifications

    Formula Weight 236.08
    Boiling Point 216° to 217°C
    Quantity 1g
    Molecular Formula C6H6BrNO2S
    MDL Number MFCD00463837
    CAS 41731-83-3
    Synonym 2-Bromothiazole-5-carboxylic acid ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 110°C (230°F)
    Chemical Name or Material Ethyl 2-bromothiazole-5-carboxylate
    Assay 98%

    Description

    Applications
    Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate

    Solubility
    Miscible is water.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit