Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-bromovalerate, 97%

Click to view available options
Quantity:
25g
100g
615-83-8
C7H13BrO2
209.083
MFCD00000159
ORSIRXYHFPHWTN-UHFFFAOYSA-N
ethyl 2-bromovalerate,ethyl2-bromovalerate,ethyl .alpha.-bromovalerate,pentanoic acid, 2-bromo-, ethyl ester,2-bromovaleric acid ethyl ester,2-bromopentanoic acid ethyl ester,2-bromo-pentanoic acid ethyl ester,pentanoic acid, bromo-, ethyl ester,ethyl bromovalerate,ethyl-2bromovalerate
79071
ethyl 2-bromopentanoate
CCCC(C(=O)OCC)Br
615-83-8
C7H13BrO2
209.083
MFCD00000159
ORSIRXYHFPHWTN-UHFFFAOYSA-N
ethyl 2-bromovalerate,ethyl2-bromovalerate,ethyl .alpha.-bromovalerate,pentanoic acid, 2-bromo-, ethyl ester,2-bromovaleric acid ethyl ester,2-bromopentanoic acid ethyl ester,2-bromo-pentanoic acid ethyl ester,pentanoic acid, bromo-, ethyl ester,ethyl bromovalerate,ethyl-2bromovalerate
79071
ethyl 2-bromopentanoate
CCCC(C(=O)OCC)Br
Quantity 25g
Flash Point 77°C (170°F)
Boiling Point 190°C to 192°C
UN Number UN3265
Beilstein 1098901
Formula Weight 209.09
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromovalerate

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.