Alfa Aesar™ Ethyl 2-butynoate, 98%

Manufacturer: Alfa Aesar™ B2537006

Quantity 5g

 View more versions of this product

Catalog No. AAB2537006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4341-76-8
    Molecular Formula C6H8O2
    Linear Formula CH3C≡CCO2CH2CH3
    Formula Weight 112.13
    MDL Number MFCD00015182
    UN Number UN3272
    Beilstein 1744948
    Synonym 2-Butynoic acid ethyl ester; Ethyl tetrolate
    Chemical Name or Material Ethyl 2-butynoate

    Specifications

    Formula Weight 112.13
    Boiling Point 163°C
    Quantity 5g
    Flash Point 53°C (127°F)
    Odor Characteristic
    MDL Number MFCD00015182
    Synonym 2-Butynoic acid ethyl ester; Ethyl tetrolate
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Molecular Formula C6H8O2
    Linear Formula CH3C≡CCO2CH2CH3
    CAS 4341-76-8
    UN Number UN3272
    Beilstein 1744948
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Ethyl 2-butynoate
    Assay 98%

    Description

    Applications
    Ethyl 2-butynoate is used as a precursor for the synthesis of 3-ethyl, 1-methyl 1-(2-nitrophenyl)-cyclopent-3-ene-1,3-dicarboxylate, alkenylsilanols and tricyclic aziridine derivatives. It acts as a methanogenesis inhibitor. As an electron deficient alkyne derivative, it plays a vital role in the preparation of 1,3-dienes catalyzed by rhodium(I)/H8-BINAP complex.

    Solubility
    Miscible with water, chloroform and ethyl acetate.

    Notes
    Air sensitive. Incompatible with oxidizing agents, bases and acids.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit