Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-chloro-3-pyridylglyoxylate, 97%

Manufacturer:  Alfa Aesar™ H5015303

Quantity 1g

 View more versions of this product

Catalog No. AAH5015303


  Check Availability
Add to cart

Specifications

Description

Applications
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

902837-56-3
213.62
2-Chloro-3-pyridylglyoxylic acid ethyl ester; Ethyl (2-chloro-3-pyridinyl)oxoacetate
C9H8ClNO3
MFCD07781194
Ethyl 2-chloro-3-pyridylglyoxylate

Specifications

902837-56-3
213.62
2-Chloro-3-pyridylglyoxylic acid ethyl ester; Ethyl (2-chloro-3-pyridinyl)oxoacetate
Ethyl 2-chloro-3-pyridylglyoxylate
C9H8ClNO3
MFCD07781194
1g
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit