Alfa Aesar™ Ethyl 2-chloro-5-pyridylglyoxylate, 97%

Manufacturer: Alfa Aesar™ H5015706

Quantity 5g

 View more versions of this product

Catalog No. AAH5015706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 902837-55-2
    Molecular Formula C9H8ClNO3
    Formula Weight 213.62
    MDL Number MFCD07781188
    Synonym 6-Chloro-3-pyridylglyoxylic acid ethyl ester; Ethyl (6-chloro-3-pyridinyl)oxoacetate
    Chemical Name or Material Ethyl 2-chloro-5-pyridylglyoxylate

    Specifications

    Formula Weight 213.62
    Molecular Formula C9H8ClNO3
    MDL Number MFCD07781188
    CAS 902837-55-2
    Synonym 6-Chloro-3-pyridylglyoxylic acid ethyl ester; Ethyl (6-chloro-3-pyridinyl)oxoacetate
    Quantity 5g
    Chemical Name or Material Ethyl 2-chloro-5-pyridylglyoxylate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit