Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloroacetoacetate, 90%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
609-15-4
C6H9ClO3
164.59
MFCD00009141
RDULEYWUGKOCMR-UHFFFAOYSA-N
ethyl 2-chloroacetoacetate,2-chloroacetoacetic acid ethyl ester,butanoic acid, 2-chloro-3-oxo-, ethyl ester,ethyl alpha-chloroacetoacetate,ethyl-2-chloracetoacetate,2-chloro-3-oxobutyric acid ethyl ester,ethyl2-chloroacetoacetate,2-chloro-3-oxobutanoic acid, ethyl ester,ethyl2-chloro-3-oxobutanoate,ethyl 2-chloro-3-oxobutyrate
11858
ethyl 2-chloro-3-oxobutanoate
CCOC(=O)C(C(=O)C)Cl
609-15-4
C6H9ClO3
164.59
MFCD00009141
RDULEYWUGKOCMR-UHFFFAOYSA-N
ethyl 2-chloroacetoacetate,2-chloroacetoacetic acid ethyl ester,butanoic acid, 2-chloro-3-oxo-, ethyl ester,ethyl alpha-chloroacetoacetate,ethyl-2-chloracetoacetate,2-chloro-3-oxobutyric acid ethyl ester,ethyl2-chloroacetoacetate,2-chloro-3-oxobutanoic acid, ethyl ester,ethyl2-chloro-3-oxobutanoate,ethyl 2-chloro-3-oxobutyrate
11858
ethyl 2-chloro-3-oxobutanoate
CCOC(=O)C(C(=O)C)Cl
Flash Point 54°C
Boiling Point 200°C
Melting Point -80°C
Specific Gravity 1.19
Refractive Index 1.4410 to 1.444
Packaging Glass bottle
Density 1.1900g/mL
Infrared Spectrum Authentic
Quantity 1L
Color Yellow
Assay Percent Range 88% min. (GC)
Linear Formula CH3COCH(Cl)CO2C2H5
Solubility Information Solubility in water: 17g/L (20°C)
Formula Weight 164.59
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Ethyl 2-chloroacetoacetate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-180-4