Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-chloropyrimidine-5-carboxylate, 97%

Click to view available options
Quantity:
250mg
1g
5g
89793-12-4
C7H7ClN2O2
186.595
MFCD09863164
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid ethyl ester,2-chloro-5-ethoxycarbonyl pyrimidine,ethyl-2-chloropyrimidine-5-carboxylate,ethyl 2-chloro-pyrimidine-5-carboxylate,5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester,ethyl2-chloropyrimidine-5-carboxylate,acmc-209r2w,ksc663i8r,ethyl 2-chloranylpyrimidine-5-carboxylate,2-chloro-5-pyrimidinecarboxylic acid ethyl ester
10487815
ethyl 2-chloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1)Cl
89793-12-4
C7H7ClN2O2
186.595
MFCD09863164
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid ethyl ester,2-chloro-5-ethoxycarbonyl pyrimidine,ethyl-2-chloropyrimidine-5-carboxylate,ethyl 2-chloro-pyrimidine-5-carboxylate,5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester,ethyl2-chloropyrimidine-5-carboxylate,acmc-209r2w,ksc663i8r,ethyl 2-chloranylpyrimidine-5-carboxylate,2-chloro-5-pyrimidinecarboxylic acid ethyl ester
10487815
ethyl 2-chloropyrimidine-5-carboxylate
CCOC(=O)C1=CN=C(N=C1)Cl
Quantity 250mg
Melting Point 52°C to 60°C
Boiling Point 80°C (760mmHg)
Beilstein 142651
Formula Weight 186.6
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloropyrimidine-5-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C