Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-cyanobutyrate, 97%

Click to view available options
Quantity:
250mg
1g
1619-58-5
C7H11NO2
141.17
MFCD02258598
FCISHUHNFYJJDU-UHFFFAOYSA-N
ethyl2-cyanobutanoate,butanoic acid, 2-cyano-, ethyl ester,ethyl 2-cyanobutyrate,ksc531a2r,2-cyanobutyric acid ethyl ester,2-cyanobutanoic acid ethyl ester,2-cyano-butyric acid ethyl ester,butanoic acid,2-cyano-, ethyl ester,2-cyanobutyric acid, ethyl ester
219611
ethyl 2-cyanobutanoate
CCC(C#N)C(=O)OCC
1619-58-5
C7H11NO2
141.17
MFCD02258598
FCISHUHNFYJJDU-UHFFFAOYSA-N
ethyl2-cyanobutanoate,butanoic acid, 2-cyano-, ethyl ester,ethyl 2-cyanobutyrate,ksc531a2r,2-cyanobutyric acid ethyl ester,2-cyanobutanoic acid ethyl ester,2-cyano-butyric acid ethyl ester,butanoic acid,2-cyano-, ethyl ester,2-cyanobutyric acid, ethyl ester
219611
ethyl 2-cyanobutanoate
CCC(C#N)C(=O)OCC
Refractive Index 1.4170
Density 0.985g/mL
Quantity 1g
Color Colorless
UN Number UN3276
Beilstein 385926
Formula Weight 141.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 2-cyanobutyrate

GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.