Alfa Aesar™ Ethyl 2-ethylacetoacetate, 95%

Manufacturer: Alfa Aesar™ B2229522

Quantity 100g

 View more versions of this product

Catalog No. AAB2229522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 607-97-6
    Molecular Formula C8H14O3
    Formula Weight 158.2
    MDL Number MFCD00039898
    Beilstein 636286
    Synonym 2-Ethylacetoacetic acid ethyl ester
    Chemical Name or Material Ethyl 2-ethylacetoacetate

    Specifications

    Formula Weight 158.2
    TSCA TSCA
    Molecular Formula C8H14O3
    Beilstein 636286
    Boiling Point 83° to 85°C (10mmHg)
    Quantity 100g
    Flash Point 77°C (170°F)
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00039898
    CAS 607-97-6
    Synonym 2-Ethylacetoacetic acid ethyl ester
    Health Hazard 1 Warning
    Chemical Name or Material Ethyl 2-ethylacetoacetate
    Assay 95%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit