Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-ethylacetoacetate, 95%

Click to view available options
Quantity:
25g
100g
607-97-6
C8H14O3
158.197
MFCD00039898
OKANYBNORCUPKZ-UHFFFAOYSA-N
ethyl 2-ethylacetoacetate,ethyl 2-acetylbutyrate,butanoic acid, 2-ethyl-3-oxo-, ethyl ester,2-ethylacetoacetic acid ethyl ester,ethyl 2-ethyl-3-ketobutyrate,ethyl alpha-ethylacetoacetate,ethyl-2-ethylacetoacetate,2-ethyl-3-oxo-butyric acid ethyl ester,ethyl .alpha.-acetylbutyrate,ethyl 2-ethyl-3-oxo-butanoate
79065
ethyl 2-ethyl-3-oxobutanoate
CCC(C(=O)C)C(=O)OCC
607-97-6
C8H14O3
158.197
MFCD00039898
OKANYBNORCUPKZ-UHFFFAOYSA-N
ethyl 2-ethylacetoacetate,ethyl 2-acetylbutyrate,butanoic acid, 2-ethyl-3-oxo-, ethyl ester,2-ethylacetoacetic acid ethyl ester,ethyl 2-ethyl-3-ketobutyrate,ethyl alpha-ethylacetoacetate,ethyl-2-ethylacetoacetate,2-ethyl-3-oxo-butyric acid ethyl ester,ethyl .alpha.-acetylbutyrate,ethyl 2-ethyl-3-oxo-butanoate
79065
ethyl 2-ethyl-3-oxobutanoate
CCC(C(=O)C)C(=O)OCC
Boiling Point 83°C to 85°C (10mmHg)
Flash Point 77°C (170°F)
Quantity 25g
Beilstein 636286
Formula Weight 158.2
Percent Purity 95%
Chemical Name or Material Ethyl 2-ethylacetoacetate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA