Alfa Aesar™ Ethyl 2-ethylbutyrate, 99%

Manufacturer: Alfa Aesar™ H2578806

Quantity 5g

 View more versions of this product

Catalog No. AAH2578806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2983-38-2
    Molecular Formula (CH3CH2)2CHCO2CH2CH3
    Formula Weight 144.21
    MDL Number MFCD00059396
    UN Number UN3272
    Synonym 2-Ethylbutyric acid ethyl ester; Ethyl diethylacetate
    Chemical Name or Material Ethyl 2-ethylbutyrate

    Specifications

    Formula Weight 144.21
    Molecular Formula (CH3CH2)2CHCO2CH2CH3
    Melting Point 149° to 152°C
    MDL Number MFCD00059396
    UN Number UN3272
    Synonym 2-Ethylbutyric acid ethyl ester; Ethyl diethylacetate
    Quantity 5g
    Flash Point 39°C (102°F)
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 2983-38-2
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Ethyl 2-ethylbutyrate
    Assay 99%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit