Alfa Aesar™ Ethyl 2-fluorobenzoate, 98+%

Manufacturer: Alfa Aesar™ A1656414

Quantity 25g

 View more versions of this product

Catalog No. AAA1656414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 443-26-5
    Molecular Formula C9H9FO2
    Formula Weight 168.17
    MDL Number MFCD00039214
    Beilstein 2084296
    Synonym 2-Fluorobenzoic acid ethyl ester
    Chemical Name or Material Ethyl 2-fluorobenzoate

    Specifications

    Formula Weight 168.17
    TSCA TSCA
    Molecular Formula C9H9FO2
    MDL Number MFCD00039214
    CAS 443-26-5
    Beilstein 2084296
    Synonym 2-Fluorobenzoic acid ethyl ester
    Boiling Point 217° to 220°C
    Quantity 25g
    Chemical Name or Material Ethyl 2-fluorobenzoate
    Assay ≥98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit