Alfa Aesar™ Ethyl 2-fluoroisobutyrate, 97%

Manufacturer: Alfa Aesar™ H3293103

Quantity 1g

 View more versions of this product

Catalog No. AAH3293103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 55816-69-8
    Molecular Formula C6H11FO2
    Formula Weight 134.15
    MDL Number MFCD08062432
    UN Number UN1992
    Synonym Ethyl 2-fluoro-2-methylpropionate; 2-Fluoroisobutyric acid ethyl ester
    Chemical Name or Material Ethyl 2-fluoroisobutyrate

    Specifications

    Formula Weight 134.15
    Molecular Formula C6H11FO2
    MDL Number MFCD08062432
    Boiling Point 122° to 126°C
    Quantity 1g
    Health Hazard 3 GHS P Statement
    P210-P280h-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    CAS 55816-69-8
    UN Number UN1992
    Synonym Ethyl 2-fluoro-2-methylpropionate; 2-Fluoroisobutyric acid ethyl ester
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H226
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Flammable liquid and vapor.
    Chemical Name or Material Ethyl 2-fluoroisobutyrate
    Assay 97%

    Description

    Applications
    Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H311-H331-H226
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280h-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit