Alfa Aesar™ Ethyl 2-hydroxyisobutyrate, 98%

Manufacturer: Alfa Aesar™ B2359236

Quantity 500g

 View more versions of this product

Catalog No. AAB2359236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80-55-7
    Molecular Formula C6H12O3
    Formula Weight 132.16
    MDL Number MFCD00004458
    UN Number UN3272
    Beilstein 878308
    Synonym Ethyl 2-hydroxy-2-methylpropionate; Ethyl 2-methyllactate
    Chemical Name or Material Ethyl 2-hydroxyisobutyrate

    Specifications

    Formula Weight 132.16
    Molecular Formula C6H12O3
    MDL Number MFCD00004458
    UN Number UN3272
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 149° to 150°C
    Quantity 500g
    Flash Point 44°C (111°F)
    RTECS Number OD5672000
    CAS 80-55-7
    Beilstein 878308
    Synonym Ethyl 2-hydroxy-2-methylpropionate; Ethyl 2-methyllactate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H319-H335
    Flammable liquid and vapour.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Ethyl 2-hydroxyisobutyrate
    Assay 98%

    GHS H Statement
    H226-H319-H335
    Flammable liquid and vapour.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit