Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-hydroxyisobutyrate, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
500g
80-55-7
C6H12O3
132.159
MFCD00004458
GFUIDHWFLMPAGY-UHFFFAOYSA-N
ethyl 2-hydroxyisobutyrate,ethyl 2-hydroxy-2-methylpropionate,ethyl 2-methyllactate,propanoic acid, 2-hydroxy-2-methyl-, ethyl ester,ethyl alpha-hydroxyisobutyrate,2-methyllactic acid ethyl ester,ethyl,a-hydroxyisobutyrate,lactic acid, 2-methyl-, ethyl ester,unii-s1530mcv3x,ethyl-2-hydroxyisobutyrate
6653
ethyl 2-hydroxy-2-methylpropanoate
CCOC(=O)C(C)(C)O
80-55-7
C6H12O3
132.159
MFCD00004458
GFUIDHWFLMPAGY-UHFFFAOYSA-N
ethyl 2-hydroxyisobutyrate,ethyl 2-hydroxy-2-methylpropionate,ethyl 2-methyllactate,propanoic acid, 2-hydroxy-2-methyl-, ethyl ester,ethyl alpha-hydroxyisobutyrate,2-methyllactic acid ethyl ester,ethyl,a-hydroxyisobutyrate,lactic acid, 2-methyl-, ethyl ester,unii-s1530mcv3x,ethyl-2-hydroxyisobutyrate
6653
ethyl 2-hydroxy-2-methylpropanoate
CCOC(=O)C(C)(C)O
Boiling Point 149°C to 150°C
Flash Point 44°C (111°F)
Quantity 25g
UN Number UN3272
Beilstein 878308
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material Ethyl 2-hydroxyisobutyrate

GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

RTECSNumber : OD5672000