Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-methylbenzoate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
87-24-1
C10H12O2
164.204
MFCD00009112
SOUAXOGPALPTTC-UHFFFAOYSA-N
ethyl o-toluate,ethyl o-methylbenzoate,ethyl orthotoluate,2-methylbenzoic acid ethyl ester,benzoic acid, 2-methyl-, ethyl ester,ethyl-o-toluate,2-methyl-benzoic acid ethyl ester,o-toluic acid ethyl ester,ethyl-2-methylbenzoate,ethylo-toluate
66598
ethyl 2-methylbenzoate
CCOC(=O)C1=CC=CC=C1C
87-24-1
C10H12O2
164.204
MFCD00009112
SOUAXOGPALPTTC-UHFFFAOYSA-N
ethyl o-toluate,ethyl o-methylbenzoate,ethyl orthotoluate,2-methylbenzoic acid ethyl ester,benzoic acid, 2-methyl-, ethyl ester,ethyl-o-toluate,2-methyl-benzoic acid ethyl ester,o-toluic acid ethyl ester,ethyl-2-methylbenzoate,ethylo-toluate
66598
ethyl 2-methylbenzoate
CCOC(=O)C1=CC=CC=C1C
Refractive Index 1.5070 to 1.5090 (20°C, 589nm)
Density 1.0320g/mL
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow
Assay Percent Range 99%
Linear Formula H3CC6H4COOC2H5
Formula Weight 164.2
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 2-methylbenzoate, 99%
Show More Show Less