Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl (2-methylphenoxy)acetate, 99%

Click to view available options
Quantity:
5g
25g
100g
93917-68-1
C11H14O3
194.23
MFCD00216559
NHFQSFIBSJPLIW-UHFFFAOYSA-N
ethyl 2-methylphenoxy acetate,ethyl 2-2-methylphenoxy acetate,ethyl 2-o-tolyloxy acetate,ethyl 2-methylphenoxyacetate,ethyl-2-methylphenoxy acetate,o-tolyloxyacetic acid ethyl ester,o-tolyloxy-acetic acid ethyl ester,ethyl 2-2-methylphenoxy ethanoate
3022879
ethyl 2-(2-methylphenoxy)acetate
CCOC(=O)COC1=CC=CC=C1C
93917-68-1
C11H14O3
194.23
MFCD00216559
NHFQSFIBSJPLIW-UHFFFAOYSA-N
ethyl 2-methylphenoxy acetate,ethyl 2-2-methylphenoxy acetate,ethyl 2-o-tolyloxy acetate,ethyl 2-methylphenoxyacetate,ethyl-2-methylphenoxy acetate,o-tolyloxyacetic acid ethyl ester,o-tolyloxy-acetic acid ethyl ester,ethyl 2-2-methylphenoxy ethanoate
3022879
ethyl 2-(2-methylphenoxy)acetate
CCOC(=O)COC1=CC=CC=C1C
Flash Point >110°C (230°F)
Quantity 5g
Boiling Point 119°C to 120°C
Formula Weight 194.23
Percent Purity 99%
Chemical Name or Material Ethyl (2-methylphenoxy)acetate