Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ethyl (2-methylphenoxy)acetate, 99%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Refractive Index | 1.503 |
Density | 1.073 |
Flash Point | >110°C (230°F) |
Quantity | 100g |
Boiling Point | 119°C to 120°C |
Formula Weight | 194.23 |
Percent Purity | 99% |
Chemical Name or Material | Ethyl (2-methylphenoxy)acetate |
Safety and Handling
EINECSNumber : 299-846-3
TSCA : No
Recommended Storage : Ambient temperatures