Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-octynoate, 98%

Click to view available options
Quantity:
5g
25g
100g
10519-20-7
C10H16O2
168.236
MFCD00015286
QPMDWYXUSMRVKT-UHFFFAOYSA-N
ethyl 2-octynoate,2-octynoic acid, ethyl ester,2-octynoic acid ethyl ester,ethyl 2-octynate,ethyl heptynecarbonate,unii-xth780u0n3,ethyl heptine carbonate,ethyl n-oct-2-ynoate,ethyl heptyne carboxylate,2-octynoic acid, ethylester
66338
ethyl oct-2-ynoate
CCCCCC#CC(=O)OCC
10519-20-7
C10H16O2
168.236
MFCD00015286
QPMDWYXUSMRVKT-UHFFFAOYSA-N
ethyl 2-octynoate,2-octynoic acid, ethyl ester,2-octynoic acid ethyl ester,ethyl 2-octynate,ethyl heptynecarbonate,unii-xth780u0n3,ethyl heptine carbonate,ethyl n-oct-2-ynoate,ethyl heptyne carboxylate,2-octynoic acid, ethylester
66338
ethyl oct-2-ynoate
CCCCCC#CC(=O)OCC
Quantity 25g
Boiling Point 85°C (6mmHg)
Refractive Index 1.447
Density 0.91
Flash Point 99°C (210°F)
Linear Formula CH3(CH2)4C≡CCO2CH2CH3
Beilstein 1759822
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl 2-octynoate

EINECSNumber : 234-058-5

TSCA : Yes

Recommended Storage : Keep cold