Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-oxobutyrate, 90%

Click to view available options
Quantity:
1g
5g
15933-07-0
C6H10O3
130.143
MFCD03265336
FJAKCEHATXBFJT-UHFFFAOYSA-N
butanoic acid, 2-oxo-, ethyl ester,ethyl 2-ketobutyrate,2-oxo-butyric acid ethyl ester,ethyl 3-methylpyruvate,ethyl a-oxobutyrate,ethyl2-oxobutanoate,ethyl 2-oxobutyrate,ethyl a-ketobutyrate,pubchem24313,ethyl-.alpha.-ketobutyrate
85195
ethyl 2-oxobutanoate
CCC(=O)C(=O)OCC
15933-07-0
C6H10O3
130.143
MFCD03265336
FJAKCEHATXBFJT-UHFFFAOYSA-N
butanoic acid, 2-oxo-, ethyl ester,ethyl 2-ketobutyrate,2-oxo-butyric acid ethyl ester,ethyl 3-methylpyruvate,ethyl a-oxobutyrate,ethyl2-oxobutanoate,ethyl 2-oxobutyrate,ethyl a-ketobutyrate,pubchem24313,ethyl-.alpha.-ketobutyrate
85195
ethyl 2-oxobutanoate
CCC(=O)C(=O)OCC
Color Yellow
Quantity 5g
Beilstein 1753936
Formula Weight 130.14
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Ethyl 2-oxobutyrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C