Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-oxocyclopentanecarboxylate, 95+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
611-10-9
C8H12O3
156.18
MFCD00001412
JHZPNBKZPAWCJD-UHFFFAOYSA-N
ethyl 2-oxocyclopentanecarboxylate,2-carbethoxycyclopentanone,ethyl cyclopentanone-2-carboxylate,cyclopentanecarboxylic acid, 2-oxo-, ethyl ester,cyclopentanone-2-carboxylic acid ethyl ester,ethyl2-oxocyclopentanecarboxylate,2-ethoxycarbonyl cyclopentanone,ethyl 2-cyclopentanonecarboxylate,.alpha.-carboethoxy cyclopentanone,ethyl 2-cyclopentanone-1-carboxylate
69136
ethyl 2-oxocyclopentane-1-carboxylate
CCOC(=O)C1CCCC1=O
611-10-9
C8H12O3
156.18
MFCD00001412
JHZPNBKZPAWCJD-UHFFFAOYSA-N
ethyl 2-oxocyclopentanecarboxylate,2-carbethoxycyclopentanone,ethyl cyclopentanone-2-carboxylate,cyclopentanecarboxylic acid, 2-oxo-, ethyl ester,cyclopentanone-2-carboxylic acid ethyl ester,ethyl2-oxocyclopentanecarboxylate,2-ethoxycarbonyl cyclopentanone,ethyl 2-cyclopentanonecarboxylate,.alpha.-carboethoxy cyclopentanone,ethyl 2-cyclopentanone-1-carboxylate
69136
ethyl 2-oxocyclopentane-1-carboxylate
CCOC(=O)C1CCCC1=O
Flash Point 77°C
Specific Gravity 1.05
Refractive Index 1.4500 to 1.4540
Infrared Spectrum Authentic
Boiling Point 224.0°C to 228.0°C
Quantity 500g
Color Colorless to Yellow
Density 1.0500g/mL
Packaging Glass bottle
Assay Percent Range 95% min. (GC)
Linear Formula (O=)C5H7CO2C2H5
Beilstein 10, 597
Solubility Information Solubility in water: 3-3.5%. Other solubilities: soluble in most organic solvent
Formula Weight 156.18
Physical Form Liquid
Percent Purity 95+%
Grade Technical
Chemical Name or Material Ethyl 2-oxocyclopentanecarboxylate, tech., 90%
Show More Show Less

EINECSNumber : 210-253-0