Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-oxopyrrolidine-4-carboxylate, 96%

Click to view available options
Quantity:
250mg
1g
60298-18-2
C7H11NO3
157.169
MFCD08274999
QWMPXTNXXREMCK-UHFFFAOYSA-N
5-oxo-pyrrolidine-3-carboxylic acid ethyl ester,ethyl5-oxopyrrolidine-3-carboxylate,5-oxo-3-pyrrolidinecarboxylic acid ethyl ester,3-pyrrolidinecarboxylic acid, 5-oxo-, ethyl ester,4-ethoxycarbonyl-2-pyrrolidone,ethyl 2-oxopyrrolidine-4-carboxylate,ethyl 5-oxopyrrolidine-3-carboxylate,ethyl 2-oxopyrrolidine-4-carboxylate,ethyl 5-oxopyrrolidine-3-carbodylate,ethyl 5-oxidanylidenepyrrolidine-3-carboxylate
44890818
ethyl 5-oxopyrrolidine-3-carboxylate
CCOC(=O)C1CC(=O)NC1
60298-18-2
C7H11NO3
157.169
MFCD08274999
QWMPXTNXXREMCK-UHFFFAOYSA-N
5-oxo-pyrrolidine-3-carboxylic acid ethyl ester,ethyl5-oxopyrrolidine-3-carboxylate,5-oxo-3-pyrrolidinecarboxylic acid ethyl ester,3-pyrrolidinecarboxylic acid, 5-oxo-, ethyl ester,4-ethoxycarbonyl-2-pyrrolidone,ethyl 2-oxopyrrolidine-4-carboxylate,ethyl 5-oxopyrrolidine-3-carboxylate,ethyl 2-oxopyrrolidine-4-carboxylate,ethyl 5-oxopyrrolidine-3-carbodylate,ethyl 5-oxidanylidenepyrrolidine-3-carboxylate
44890818
ethyl 5-oxopyrrolidine-3-carboxylate
CCOC(=O)C1CC(=O)NC1
Quantity 1g
Color Brown
Melting Point 40°C to 47°C
Formula Weight 157.17
Physical Form Solid
Percent Purity 96%
Chemical Name or Material Ethyl 2-oxopyrrolidine-4-carboxylate