Alfa Aesar™ Ethyl 2-pentynyl ether, 97%

Manufacturer: Alfa Aesar™ H2596014

Quantity 25g

 View more versions of this product

Catalog No. AAH2596014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20635-10-3
    Molecular Formula C7H12O
    Linear Formula CH3CH2C≡CCH2OCH2CH3
    Formula Weight 112.17
    MDL Number MFCD00048702
    UN Number UN3271
    Synonym 1-Ethoxy-2-pentyne
    Chemical Name or Material Ethyl 2-pentynyl ether

    Specifications

    Formula Weight 112.17
    Molecular Formula C7H12O
    Linear Formula CH3CH2C≡CCH2OCH2CH3
    CAS 20635-10-3
    UN Number UN3271
    Synonym 1-Ethoxy-2-pentyne
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    Flash Point 30°C (86°F)
    Boiling Point 63° to 65°C (20mmHg)
    Quantity 25g
    MDL Number MFCD00048702
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Ethyl 2-pentynyl ether
    Assay 97%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit