Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 2-pyridineacetate, 98%

Click to view available options
Quantity:
5g
25g
100g
2739-98-2
C9H11NO2
165.192
MFCD00006359
IUDKTVXSXWAKJO-UHFFFAOYSA-N
ethyl 2-pyridylacetate,ethyl 2-pyridin-2-yl acetate,ethyl 2-pyridinylacetate,ethyl pyridine-2-acetate,2-pyridineacetic acid ethyl ester,ethyl 2-pyridineacetate,ethyl 2-2-pyridyl acetate,2-pyridylacetic acid ethyl ester,ethyl pyridin-2-ylacetate,2-pyridineacetic acid, ethyl ester
75960
ethyl 2-pyridin-2-ylacetate
CCOC(=O)CC1=CC=CC=N1
2739-98-2
C9H11NO2
165.192
MFCD00006359
IUDKTVXSXWAKJO-UHFFFAOYSA-N
ethyl 2-pyridylacetate,ethyl 2-pyridin-2-yl acetate,ethyl 2-pyridinylacetate,ethyl pyridine-2-acetate,2-pyridineacetic acid ethyl ester,ethyl 2-pyridineacetate,ethyl 2-2-pyridyl acetate,2-pyridylacetic acid ethyl ester,ethyl pyridin-2-ylacetate,2-pyridineacetic acid, ethyl ester
75960
ethyl 2-pyridin-2-ylacetate
CCOC(=O)CC1=CC=CC=N1
Applications
Ethyl 2-pyridineacetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Flash Point >110°C (230°F)
Quantity 25g
Boiling Point 69°C to 70°C (0.05mmHg)
Beilstein 123817
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Ethyl 2-pyridineacetate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning