Alfa Aesar™ Ethyl 3-(2-methoxy-3-pyridyl)acrylate

Manufacturer: Alfa Aesar™ H5011906

Quantity 5g

 View more versions of this product

Catalog No. AAH5011906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 912760-91-9
    Molecular Formula C11H13NO3
    Formula Weight 207.23
    MDL Number MFCD07781176
    Synonym 3-(2-Methoxy-3-pyridyl)acrylic acid ethyl ester
    Chemical Name or Material Ethyl 3-(2-methoxy-3-pyridyl)acrylate

    Specifications

    Formula Weight 207.23
    Molecular Formula C11H13NO3
    MDL Number MFCD07781176
    CAS 912760-91-9
    Synonym 3-(2-Methoxy-3-pyridyl)acrylic acid ethyl ester
    Quantity 5g
    Chemical Name or Material Ethyl 3-(2-methoxy-3-pyridyl)acrylate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit