Alfa Aesar™ Ethyl 3-(3,4,5-trimethoxyphenyl)propionate, 98%

Manufacturer: Alfa Aesar™ A1245906

Quantity 5g

 View more versions of this product

Catalog No. AAA1245906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 70311-20-5
    Molecular Formula C14H20O5
    Formula Weight 268.31
    MDL Number MFCD00017522
    Synonym 3-(3,4,5-Trimethoxyphenyl)propionic acid ethyl ester
    Chemical Name or Material Ethyl 3- (3,4,5-trimethoxyphenyl)propionate

    Specifications

    Formula Weight 268.31
    MDL Number MFCD00017522
    CAS 70311-20-5
    Molecular Formula C14H20O5
    Synonym 3-(3,4,5-Trimethoxyphenyl)propionic acid ethyl ester
    Quantity 5g
    Chemical Name or Material Ethyl 3- (3,4,5-trimethoxyphenyl)propionate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit