Alfa Aesar™ Ethyl 3,4-dihydroxycinnamate, 97%

Manufacturer: Alfa Aesar™ B2480814

Quantity 25g

 View more versions of this product

Catalog No. AAB2480814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 66648-50-8
    Molecular Formula C11H12O4
    Formula Weight 208.22
    MDL Number MFCD00045754
    Synonym Ethyl caffeate; 3,4-Dihydroxycinnamic acid ethyl ester
    Chemical Name or Material Ethyl 3,4-dihydroxycinnamate

    Specifications

    Formula Weight 208.22
    Molecular Formula C11H12O4
    MDL Number MFCD00045754
    CAS 66648-50-8
    Synonym Ethyl caffeate; 3,4-Dihydroxycinnamic acid ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    Chemical Name or Material Ethyl 3,4-dihydroxycinnamate
    Assay 97%

    Description

    Applications
    Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents Store at -20°C.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit