Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%

Click to view available options
Quantity:
1g
5g
25g
96617-71-9
C11H8F6O2
286.173
MFCD01320684
NBWZJMOEFTYMOF-UHFFFAOYSA-N
ethyl 3,5-bis trifluoromethyl benzoate,3,5-bis-trifluoromethyl-benzoic acid ethyl ester,3,5-bis trifluoromethyl benzoic acid ethyl ester,rarechem al bi 0207,acmc-20al07,ethyl 3?5-bis trifluoromethyl benzoate,ethyl 3 5-bis trifluoromethyl benzoate,benzoic acid, 3,5-bis trifluoromethyl-, ethyl ester
2733458
ethyl 3,5-bis(trifluoromethyl)benzoate
CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
96617-71-9
C11H8F6O2
286.173
MFCD01320684
NBWZJMOEFTYMOF-UHFFFAOYSA-N
ethyl 3,5-bis trifluoromethyl benzoate,3,5-bis-trifluoromethyl-benzoic acid ethyl ester,3,5-bis trifluoromethyl benzoic acid ethyl ester,rarechem al bi 0207,acmc-20al07,ethyl 3?5-bis trifluoromethyl benzoate,ethyl 3 5-bis trifluoromethyl benzoate,benzoic acid, 3,5-bis trifluoromethyl-, ethyl ester
2733458
ethyl 3,5-bis(trifluoromethyl)benzoate
CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 25g
Formula Weight 286.17
Percent Purity 99%
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)benzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning