Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2199-44-2
C9H13NO2
167.21
MFCD00010645
IZSBSZYFPYIJDI-UHFFFAOYSA-N
3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester,ethyl 3,5-dimethylpyrrole-2-carboxylate,ethyl 3,5-dimethyl-2-pyrrolecarboxylate,1h-pyrrole-2-carboxylic acid, 3,5-dimethyl-, ethyl ester,3,5-dimethyl-1h-pyrrole-2-carboxylic acidethyl ester,ethyl 3,5-dimethyl-1h-pyrrole-2-carboxylate,pubchem23984,acmc-1cdc7,3,5-dimethyl-1h-pyrrole-2-carboxylicacidethylester,ksc496q7t
137477
ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
CCOC(=O)C1=C(C=C(N1)C)C
2199-44-2
C9H13NO2
167.21
MFCD00010645
IZSBSZYFPYIJDI-UHFFFAOYSA-N
3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester,ethyl 3,5-dimethylpyrrole-2-carboxylate,ethyl 3,5-dimethyl-2-pyrrolecarboxylate,1h-pyrrole-2-carboxylic acid, 3,5-dimethyl-, ethyl ester,3,5-dimethyl-1h-pyrrole-2-carboxylic acidethyl ester,ethyl 3,5-dimethyl-1h-pyrrole-2-carboxylate,pubchem23984,acmc-1cdc7,3,5-dimethyl-1h-pyrrole-2-carboxylicacidethylester,ksc496q7t
137477
ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
CCOC(=O)C1=C(C=C(N1)C)C
Color Yellow
Packaging Glass bottle
Boiling Point 135°C to 136°C (10.0mmHg)
Melting Point 122°C to 126°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 167.21
Physical Form Buff Powder
Percent Purity 98%
Chemical Name or Material Ethyl 3, 5-dimethyl-1H-pyrrole-2-carboxylate, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning