Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3,5-dinitrobenzoate, 98%

Click to view available options
Quantity:
50g
250g
618-71-3
C9H8N2O6
240.171
MFCD00007232
IBQREHJPMPCXQA-UHFFFAOYSA-N
benzoic acid, 3,5-dinitro-, ethyl ester,acmc-20al01,ethyl 3,5-dinitrobenzoate,3,5-dinitrobenzoic acid ethyl ester,3,5-dinitro-benzoic acid ethyl ester,benzoic acid,5-dinitro-, ethyl ester,benzoic acid,3,5-dinitro-, ethyl ester
69255
ethyl 3,5-dinitrobenzoate
CCOC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]
618-71-3
C9H8N2O6
240.171
MFCD00007232
IBQREHJPMPCXQA-UHFFFAOYSA-N
benzoic acid, 3,5-dinitro-, ethyl ester,acmc-20al01,ethyl 3,5-dinitrobenzoate,3,5-dinitrobenzoic acid ethyl ester,3,5-dinitro-benzoic acid ethyl ester,benzoic acid,5-dinitro-, ethyl ester,benzoic acid,3,5-dinitro-, ethyl ester
69255
ethyl 3,5-dinitrobenzoate
CCOC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]
Melting Point 91°C to 92°C
Quantity 50g
Beilstein 1995473
Formula Weight 240.17
Percent Purity 98%
Chemical Name or Material Ethyl 3,5-dinitrobenzoate