Alfa Aesar™ Ethyl 3-amino-1H-pyrazole-4-carboxylate, 99%

Manufacturer: Alfa Aesar™ A1088222

Quantity 100g

 View more versions of this product

Catalog No. AAA1088222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6994-25-8
    Molecular Formula C6H9N3O2
    Formula Weight 155.16
    MDL Number MFCD00005238
    Beilstein 4964
    Synonym 3-Amino-4-carboethoxy-1H-pyrazole; 3-Amino-1H-pyrazole-4-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 3-amino-1H-pyrazole-4-carboxylate

    Specifications

    Formula Weight 155.16
    Beilstein 4964
    Synonym 3-Amino-4-carboethoxy-1H-pyrazole; 3-Amino-1H-pyrazole-4-carboxylic acid ethyl ester
    Quantity 100g
    Molecular Formula C6H9N3O2
    MDL Number MFCD00005238
    CAS 6994-25-8
    Melting Point 104° to 107°C
    Chemical Name or Material Ethyl 3-amino-1H-pyrazole-4-carboxylate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit