Alfa Aesar™ Ethyl 3-amino-4,4,4-trifluorocrotonate, 97%

Manufacturer: Alfa Aesar™ B2254514

Quantity 25g

 View more versions of this product

Catalog No. AAB2254514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 372-29-2
    Molecular Formula C6H8F3NO2
    Formula Weight 183.13
    MDL Number MFCD00068195
    UN Number UN3259
    Beilstein 4397839
    Synonym 3-Amino-4,4,4-trifluorocrotonic acid ethyl ester
    Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorocrotonate

    Specifications

    Formula Weight 183.13
    Molecular Formula C6H8F3NO2
    MDL Number MFCD00068195
    CAS 372-29-2
    UN Number UN3259
    Beilstein 4397839
    Synonym 3-Amino-4,4,4-trifluorocrotonic acid ethyl ester
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 24° to 26°C
    Odor Amine-like
    Flash Point 65°C (149°F)
    Health Hazard 1 Danger
    Boiling Point 83° to 85°C (15mmHg)
    Quantity 25g
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorocrotonate
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit