Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-aminocrotonate, 98.5%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
7318-00-5
C6H11NO2
129.16
MFCD02730138
YPMPTULBFPFSEQ-PLNGDYQASA-N
z-ethyl 3-aminobut-2-enoate,ethyl 3-aminocrotonate,ethyl 3-aminobut-2-enoate,ethyl 2z-3-aminobut-2-enoate,2-butenoic acid, 3-amino-, ethyl ester, 2z,ethyl z-3-aminobut-2-enoate,ethyl 3-amino-2-butenoate,.beta.-aminocrotonic acid ethyl ester,2-butenoic acid, 3-amino-, ethyl ester,ethyl .beta.-aminocrotonate
643756
ethyl (Z)-3-aminobut-2-enoate
CCOC(=O)C=C(C)N
7318-00-5
C6H11NO2
129.16
MFCD02730138
YPMPTULBFPFSEQ-PLNGDYQASA-N
z-ethyl 3-aminobut-2-enoate,ethyl 3-aminocrotonate,ethyl 3-aminobut-2-enoate,ethyl 2z-3-aminobut-2-enoate,2-butenoic acid, 3-amino-, ethyl ester, 2z,ethyl z-3-aminobut-2-enoate,ethyl 3-amino-2-butenoate,.beta.-aminocrotonic acid ethyl ester,2-butenoic acid, 3-amino-, ethyl ester,ethyl .beta.-aminocrotonate
643756
ethyl (Z)-3-aminobut-2-enoate
CCOC(=O)C=C(C)N
Color Yellow
Infrared Spectrum Authentic
Density 1.0220g/mL
Packaging Glass bottle
Refractive Index 1.5000 to 1.5020
Specific Gravity 1.022
Boiling Point 210.0°C to 215.0°C
Flash Point 97°C
Quantity 500g
Melting Point 33.0°C to 35.0°C
Assay Percent Range 98.5%
Linear Formula CH3C(NH2)=CHCO2C2H5
Beilstein 03, 654
Formula Weight 129.16
Physical Form Low Melting Solid or Liquid
Percent Purity 98.5%
Chemical Name or Material Ethyl 3-aminocrotonate, 98.5%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 230-782-0