Learn More
Alfa Aesar™ Ethyl 3-bromobenzoate, 98+%
Manufacturer:  Alfa Aesar™ A1025130
Quantity 250g
 View more versions of this product
Catalog No. AAA1025130
  Check Availability
Add to cart
Specifications
Description
Applications
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4 ft.-methyl-biphenyl-3-carboxylic acid ethyl ester.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Description & Specifications
Chemical Identifiers
24398-88-7
229.07
1867121
Ethyl 3-bromobenzoate
C9H9BrO2
MFCD00013529
3-Bromobenzoic acid ethyl ester
Specifications
24398-88-7
C9H9BrO2
>110°C (230°F)
MFCD00013529
3-Bromobenzoic acid ethyl ester
≥98%
261°C
250g
229.07
1867121
Ethyl 3-bromobenzoate