Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-chlorophenylacetate, +98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
14062-29-4
C10H11ClO2
198.646
MFCD07787498
SLNHJHHDMDGTGB-UHFFFAOYSA-N
ethyl 2-3-chlorophenyl acetate,ethyl 3-chlorophenyl acetate,3-chloro-phenyl-acetic acid ethyl ester,ethyl m-chlorophenylacetate,3-chlorophenyl acetic acid ethyl ester,m-chlorophenylacetic acid ethyl ester,benzeneacetic acid, 3-chloro-, ethyl ester,ethyl 3-chlorophenylacetate,3-chlorobenzeneacetic acid ethyl ester
8063366
ethyl 2-(3-chlorophenyl)acetate
CCOC(=O)CC1=CC(=CC=C1)Cl
14062-29-4
C10H11ClO2
198.646
MFCD07787498
SLNHJHHDMDGTGB-UHFFFAOYSA-N
ethyl 2-3-chlorophenyl acetate,ethyl 3-chlorophenyl acetate,3-chloro-phenyl-acetic acid ethyl ester,ethyl m-chlorophenylacetate,3-chlorophenyl acetic acid ethyl ester,m-chlorophenylacetic acid ethyl ester,benzeneacetic acid, 3-chloro-, ethyl ester,ethyl 3-chlorophenylacetate,3-chlorobenzeneacetic acid ethyl ester
8063366
ethyl 2-(3-chlorophenyl)acetate
CCOC(=O)CC1=CC(=CC=C1)Cl
Quantity 25g
Packaging Glass bottle
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98+%
Linear Formula ClC6H4CH2(O)OCH2CH3
Formula Weight 198.65
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material Ethyl 3-chlorophenylacetate
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.