Alfa Aesar™ Ethyl 3-chloropropionate, 98%

Manufacturer: Alfa Aesar™ B2390714

Quantity 25g

 View more versions of this product

Catalog No. AAB2390714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 623-71-2
    Molecular Formula ClCH2CH2CO2CH2CH3
    Formula Weight 136.58
    MDL Number MFCD00000989
    Beilstein 385698
    Synonym 3-Chloropropionic acid ethyl ester
    Chemical Name or Material Ethyl 3-chloropropionate

    Specifications

    Formula Weight 136.58
    Molecular Formula ClCH2CH2CO2CH2CH3
    MDL Number MFCD00000989
    Beilstein 385698
    Health Hazard 1 Warning
    Boiling Point 162° to 164°C
    Quantity 25g
    Flash Point 61°C (142°F)
    TSCA TSCA
    CAS 623-71-2
    Synonym 3-Chloropropionic acid ethyl ester
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Ethyl 3-chloropropionate
    Assay 98%

    Description

    Applications
    It can be used to produce (1-ethoxycyclopropyloxy)trimethylsilane by heating.

    Solubility
    Immiscible in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit