Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-chloropropionate, 98%

Manufacturer:  Alfa Aesar™ B2390730

Quantity 250g

 View more versions of this product

Catalog No. AAB2390730


  Check Availability
Add to cart

Specifications

Description

Applications
It can be used to produce (1-ethoxycyclopropyloxy)trimethylsilane by heating.

Solubility
Immiscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

623-71-2
136.58
385698
Ethyl 3-chloropropionate
ClCH2CH2CO2CH2CH3
MFCD00000989
3-Chloropropionic acid ethyl ester

Specifications

623-71-2
136.58
385698
162° to 164°C
61°C (142°F)
98%
ClCH2CH2CO2CH2CH3
MFCD00000989
3-Chloropropionic acid ethyl ester
250g
Ethyl 3-chloropropionate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit