Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl 3-(dimethylamino)acrylate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water 90 g/L (25°C).
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water 90 g/L (25°C).
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 17°C to 18°C |
Boiling Point | 118°C to 121°C (10 mmHg) |
Refractive Index | 1.512 |
Linear Formula | (CH3)2NCH=CHCO2CH2CH3 |
Quantity | 10 g |
Solubility Information | Soluble in water 90g/L (25°C). |
Formula Weight | 143.19 |
Percent Purity | 99% |
Chemical Name or Material | Ethyl 3-(dimethylamino)acrylate |
Safety and Handling
Hazard Category | H317 |
Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
EINECSNumber | 402-650-4 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction