Alfa Aesar™ Ethyl 3-ethoxy-2-butenoate, 97%

Manufacturer: Alfa Aesar™ A1109714

Quantity 25g

 View more versions of this product

Catalog No. AAA1109714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 998-91-4
    Molecular Formula C8H14O3
    Formula Weight 158.2
    MDL Number MFCD00026932
    Beilstein 1757006
    Synonym 3-Ethoxy-2-butenoic acid ethyl ester; Ethyl 3-ethoxycrotonate
    Chemical Name or Material Ethyl 3-ethoxy-2-butenoate

    Specifications

    Formula Weight 158.2
    CAS 998-91-4
    Beilstein 1757006
    MDL Number MFCD00026932
    Synonym 3-Ethoxy-2-butenoic acid ethyl ester; Ethyl 3-ethoxycrotonate
    Melting Point 28° to 32°C
    Quantity 25g
    Molecular Formula C8H14O3
    Boiling Point 86° to 88°C (14mmHg)
    Chemical Name or Material Ethyl 3-ethoxy-2-butenoate
    Assay 97%

    Description

    Applications
    Ethyl 3-ethoxy-2-butenoate is used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit