Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-ethoxypropionate, 99+%

Click to view available options
Quantity:
1000mL
763-69-9
C7H14O3
146.186
MFCD00051356
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
ethyl 3-ethoxypropionate,3-ethoxypropionic acid ethyl ester,propanoic acid, 3-ethoxy-, ethyl ester,eep solvent,ethoxypropionic acid, ethyl ester,ethyl beta-ethoxypropionate,unii-ec38rsj79j,propionic acid, 3-ethoxy-, ethyl ester,ethyl-3-ethoxypropionate,ethylester kyseliny 3-ethoxypropionove
12989
ethyl 3-ethoxypropanoate
CCOCCC(=O)OCC
763-69-9
C7H14O3
146.186
MFCD00051356
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
ethyl 3-ethoxypropionate,3-ethoxypropionic acid ethyl ester,propanoic acid, 3-ethoxy-, ethyl ester,eep solvent,ethoxypropionic acid, ethyl ester,ethyl beta-ethoxypropionate,unii-ec38rsj79j,propionic acid, 3-ethoxy-, ethyl ester,ethyl-3-ethoxypropionate,ethylester kyseliny 3-ethoxypropionove
12989
ethyl 3-ethoxypropanoate
CCOCCC(=O)OCC
Applications
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

Solubility
Miscible with most organic solvents. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents, metals and strong acids.
Melting Point -100°C
Odor Ester-like
Quantity 1000mL
Boiling Point 169°C to 171°C
Flash Point 59°C (142°F)
Linear Formula CH3CH2OCH2CH2CO2CH2CH3
UN Number UN3272
Formula Weight 146.19
Percent Purity ≥99%
Chemical Name or Material Ethyl 3-ethoxypropionate

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

RTECSNumber : UF3325000

TSCA : TSCA