Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-ethoxypropionate, 99+%

Manufacturer:  Alfa Aesar™ B24024AK

Quantity 250mL

 View more versions of this product

Catalog No. AAB24024AK


  Check Availability
Add to cart

Specifications

Description

Applications
Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.

Solubility
Miscible with most organic solvents. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents, metals and strong acids.
Description & Specifications

Chemical Identifiers

763-69-9
CH3CH2OCH2CH2CO2CH2CH3
MFCD00051356
3-Ethoxypropionic acid ethyl ester
C7H14O3
146.19
UN3272
Ethyl 3-ethoxypropionate

Specifications

763-69-9
CH3CH2OCH2CH2CO2CH2CH3
MFCD00051356
3-Ethoxypropionic acid ethyl ester
−100°C
250mL
Ethyl 3-ethoxypropionate
C7H14O3
146.19
UN3272
169° to 171°C
Ester-like
59°C (142°F)
≥99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

RTECSNumber : UF3325000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit