Alfa Aesar™ Ethyl 3-(furfurylthio)propionate, 97%

Manufacturer: Alfa Aesar™ B2176618

Quantity 50g

 View more versions of this product

Catalog No. AAB2176618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94278-27-0
    Molecular Formula C10H14O3S
    Formula Weight 214.29
    MDL Number MFCD00051819
    Beilstein 4311671
    Synonym Ethyl 3-(furfurylmercapto)propionate; 3-(Furfurylthio)propionic acid ethyl ester
    Chemical Name or Material Ethyl 3-(furfurylthio)propionate

    Specifications

    Formula Weight 214.29
    Molecular Formula C10H14O3S
    MDL Number MFCD00051819
    CAS 94278-27-0
    Beilstein 4311671
    Synonym Ethyl 3-(furfurylmercapto)propionate; 3-(Furfurylthio)propionic acid ethyl ester
    Boiling Point 120° to 122°C (0.5mmHg)
    Quantity 50g
    Flash Point 99°C (210°F)
    Chemical Name or Material Ethyl 3-(furfurylthio)propionate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit