Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 3-hydroxy-3-phenylpropionate, 95%

Click to view available options
Quantity:
250mg
1g
5g
5764-85-2
C11H14O3
194.23
MFCD00086891
DVIBDQWVFHDBOP-UHFFFAOYSA-N
ethyl 3-hydroxy-3-phenylpropionate,3-hydroxy-3-phenylpropionic acid ethyl ester,hydracrylic acid, 3-phenyl-, ethyl ester,3-hydroxy-3-phenyl-propionic acid ethyl ester,benzenepropanoic acid, .beta.-hydroxy-, ethyl ester,3-hydroxy-3-phenyl-propionicacidethylester,ethyl-3-hydroxy-3-phenyl propionate,ethyl 3-hydroxy-3-phenyl-propanoate,ethyl 3-phenylhydracrylate, +/-,acmc-20apru
99818
ethyl 3-hydroxy-3-phenylpropanoate
CCOC(=O)CC(C1=CC=CC=C1)O
5764-85-2
C11H14O3
194.23
MFCD00086891
DVIBDQWVFHDBOP-UHFFFAOYSA-N
ethyl 3-hydroxy-3-phenylpropionate,3-hydroxy-3-phenylpropionic acid ethyl ester,hydracrylic acid, 3-phenyl-, ethyl ester,3-hydroxy-3-phenyl-propionic acid ethyl ester,benzenepropanoic acid, .beta.-hydroxy-, ethyl ester,3-hydroxy-3-phenyl-propionicacidethylester,ethyl-3-hydroxy-3-phenyl propionate,ethyl 3-hydroxy-3-phenyl-propanoate,ethyl 3-phenylhydracrylate, +/-,acmc-20apru
99818
ethyl 3-hydroxy-3-phenylpropanoate
CCOC(=O)CC(C1=CC=CC=C1)O
Refractive Index 1.5080
Density 1.088g/mL
Quantity 5g
Beilstein 2211602
Formula Weight 194.23
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Ethyl 3-hydroxy-3-phenylpropionate