Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-indoleacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
778-82-5
C12H13NO2
203.24
MFCD00005635
HUDBDWIQSIGUDI-UHFFFAOYSA-N
ethyl 3-indoleacetate,ethyl 2-1h-indol-3-yl acetate,indole-3-acetic acid ethyl ester,ethyl indole-3-acetate,ethyl indol-3-ylacetate,1h-indole-3-acetic acid, ethyl ester,ethyl 1h-indol-3-ylacetate,indole-3-acetic acid, ethyl ester,ethyl beta-indolylacetate,ethyl3-indoleacetate
13067
ethyl 2-(1H-indol-3-yl)acetate
CCOC(=O)CC1=CNC2=CC=CC=C21
778-82-5
C12H13NO2
203.24
MFCD00005635
HUDBDWIQSIGUDI-UHFFFAOYSA-N
ethyl 3-indoleacetate,ethyl 2-1h-indol-3-yl acetate,indole-3-acetic acid ethyl ester,ethyl indole-3-acetate,ethyl indol-3-ylacetate,1h-indole-3-acetic acid, ethyl ester,ethyl 1h-indol-3-ylacetate,indole-3-acetic acid, ethyl ester,ethyl beta-indolylacetate,ethyl3-indoleacetate
13067
ethyl 2-(1H-indol-3-yl)acetate
CCOC(=O)CC1=CNC2=CC=CC=C21
Quantity 5g
Boiling Point 164.0°C to 166.0°C (1.0mmHg)
Flash Point >110°C
Melting Point 42.0°C to 45.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone
Formula Weight 203.24
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Ethyl 3-indoleacetate, 99%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 212-296-