Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-mercaptopropionate, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
250g
Packaging:
Glass bottle
5466-06-8
C5H10O2S
134.19
CJQWLNNCQIHKHP-UHFFFAOYSA-N
hydroxylamine hydrochloride,hydroxylamine hcl,hydroxylammonium chloride,oxammonium hydrochloride,hydroxylamine, hydrochloride,hydroxyamine hydrochloride,hydroxylamine chloride 1:1,hydroxylamine, hydrochloride 1:1,hydroxyamine hcl,oxammonium hci
443297
CHEBI:5807
ethyl 3-sulfanylpropanoate
CCOC(=O)CCS
5466-06-8
C5H10O2S
134.19
CJQWLNNCQIHKHP-UHFFFAOYSA-N
hydroxylamine hydrochloride,hydroxylamine hcl,hydroxylammonium chloride,oxammonium hydrochloride,hydroxylamine, hydrochloride,hydroxyamine hydrochloride,hydroxylamine chloride 1:1,hydroxylamine, hydrochloride 1:1,hydroxyamine hcl,oxammonium hci
443297
CHEBI:5807
ethyl 3-sulfanylpropanoate
CCOC(=O)CCS
Flash Point 72°C
Infrared Spectrum Authentic
Color Colorless to Yellow
Boiling Point 75.0°C to 76.0°C (10.0mmHg)
Density 1.0590g/mL
Packaging Glass bottle
Quantity 5g
Specific Gravity 1.059
Refractive Index 1.4560 to 1.4580
Assay Percent Range 98%
Linear Formula HSCH2CH2CO2C2H5
Beilstein 03,555
Formula Weight 134.2
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl 3-mercaptopropionate, 99%
Show More Show Less

Warning

EINECSNumber : 226-771-5

TSCA : TSCA