Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Mercaptopropionate, Approx. 99%, Spectrum™ Chemical
SDP

Catalog Number 18602491
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
5466-06-8
C5H10O2S
134.19
CJQWLNNCQIHKHP-UHFFFAOYSA-N
ethyl 3-sulfanylpropanoate
CCOC(=O)CCS
5466-06-8
C5H10O2S
134.19
CJQWLNNCQIHKHP-UHFFFAOYSA-N
ethyl 3-sulfanylpropanoate
CCOC(=O)CCS

E2745, 5466-06-8, C5H10O2S

Spectrum™ Chemical Ethyl 3-Mercaptopropionate
CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity Approx. 99%
Grade Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit