Alfa Aesar™ Ethyl 3-methoxybenzoate, 98+%

Manufacturer: Alfa Aesar™ A1090218

Quantity 50g

 View more versions of this product

Catalog No. AAA1090218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10259-22-0
    Molecular Formula C10H12O3
    Formula Weight 180.2
    MDL Number MFCD00015149
    Beilstein 2576420
    Synonym Ethyl m-anisate; 3-Methoxybenzoic acid ethyl ester
    Chemical Name or Material Ethyl 3-methoxybenzoate

    Specifications

    Formula Weight 180.2
    Molecular Formula C10H12O3
    CAS 10259-22-0
    Synonym Ethyl m-anisate; 3-Methoxybenzoic acid ethyl ester
    Quantity 50g
    MDL Number MFCD00015149
    Beilstein 2576420
    Boiling Point 250° to 252°C
    Chemical Name or Material Ethyl 3-methoxybenzoate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit