Alfa Aesar™ Ethyl 3-methyl-1H-pyrazole-5-carboxylate, 97%

Manufacturer: Alfa Aesar™ A1230414

Quantity 25g

 View more versions of this product

Catalog No. AAA1230414

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 4027-57-0
    Molecular Formula C7H10N2O2
    Formula Weight 154.17
    MDL Number MFCD00052514
    Beilstein 4192
    Synonym 3-Methyl-1H-pyrazole-5-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl 3-methyl-1H-pyrazole-5-carboxylate

    Specifications

    Formula Weight 154.17
    MDL Number MFCD00052514
    CAS 4027-57-0
    Quantity 25g
    Beilstein 4192
    Synonym 3-Methyl-1H-pyrazole-5-carboxylic acid ethyl ester
    Molecular Formula C7H10N2O2
    Melting Point 80° to 83°C
    Chemical Name or Material Ethyl 3-methyl-1H-pyrazole-5-carboxylate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit