Alfa Aesar™ Ethyl 3-methyl-3-phenylglycidate, cis + trans, 98%

Manufacturer: Alfa Aesar™ B2365418

Quantity 50g

 View more versions of this product

Catalog No. AAB2365418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77-83-8
    Molecular Formula C12H14O3
    Formula Weight 206.24
    MDL Number MFCD00022338
    Beilstein 12299
    Synonym 3-Methyl-3-phenylglycidic acid ethyl ester; 3-Phenyl-2,3-epoxybutyric acid ethyl ester
    Chemical Name or Material Ethyl 3-methyl-3-phenylglycidate, cis + trans

    Specifications

    Formula Weight 206.24
    MDL Number MFCD00022338
    CAS 77-83-8
    Boiling Point 272° to 275°C
    Flash Point 134°C (273°F)
    TSCA TSCA
    Molecular Formula C12H14O3
    Beilstein 12299
    Synonym 3-Methyl-3-phenylglycidic acid ethyl ester; 3-Phenyl-2,3-epoxybutyric acid ethyl ester
    Quantity 50g
    RTECS Number MW5250000
    Chemical Name or Material Ethyl 3-methyl-3-phenylglycidate, cis + trans
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit